Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tributylmethylammonium Methyl Carbonate Solution

Catalog No. 67117710G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-117-710G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 67-117-710G Supplier Sigma Aldrich Supplier No. 67117710G
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tributylmethylammonium methyl carbonate can be used in the preparation of counter-cation anionic monomers by reacting with 2-acrylamido-2- methylpropanesulfonic acid and acrylic acid.

Specifications

CAS 53042-33-4
Density 0.882 g/mL at 25°C
Molecular Formula C2H9BLiN
Refractive Index n20/D 1.408
Linear Formula C2H9BLiN
MDL Number MFCD07784404
Quantity 100 mL
Synonym Methyltributylammonium methyl carbonate
Molecular Weight (g/mol) 64.85 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.