Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tributylphosphine

Catalog No. 90827500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-827-500ML 500 mL
90-827-100ML 100 mL
90-827-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 90-827-500ML Supplier Sigma Aldrich Supplier No. 90827500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tributylphosphine (TBP) is a potential nucleophile. It acts as a weak base in organic solvents. It is an important catalyst for the acylation reactions. It is an efficient promoting reagent for the ring-opening reaction of various epoxides and aziridines. TBP in conjugation with 1,1'sa-(azodicarbonyl)dipiperidine (ADDP) constitutes Mitsunobu reagent. This reactive phosphine allows the formation of a strained lactone.Tributylphosphine was used in the preparation of silver nanoparticles (AgNPs) functionalized with an organometallic bifunctional thiol containing Pt(II) centers. It may be used in the following processes:As reducing agent for alkyl disulfides and aromatic disulfides. As catalyst for the synthesis of 2-substituted 1,3-benzoselenazoles.As promoter for the ring opening of epoxides and aziridines with nucleophiles.As a reagent in the preparation of 6-substituted penicillanate esters by reduction of 6-bromo-6-substituted penicillanate esters in high diastereoselectivity. As a catalyst in the acylation reaction of alcohols.As a catalyst to prepare rotaxanes by the acylation of corresponding pseudorotaxanes using 3,5-dimethylbenzoic anhydride. As a catalyst to prepare vinyl thioethers by the Michael addition of ethanethiol to various alkynyl ketones.As a promoter in the conjugate addition of non-nucleophilic N-containing compounds with Michael acceptors.

Specifications

CAS 998-40-3
Density 0.81 g/mL at 25°C
Boiling Point 150°C (50 mmHg, lit.)
Molecular Formula C12H27P
Linear Formula [CH3(CH2)3]3P
Refractive Index n20/D 1.462; n20/D 1.463
Quantity 500 mL
MDL Number MFCD00009462
Synonym P(n-Bu)3; TBP
Molecular Weight (g/mol) 202.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.