Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tributyltin Chloride

Catalog No. T50202500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T50202500G 500 g
T50202100G 100 g
T502025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. T50202500G Supplier Sigma Aldrich Supplier No. T50202500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tributyltin chloride (Bu3SnCl or tri-n-butylstannane)is anorganotin compound that can be used as a reagent for: The synthesis of fluorinated stannanes by reacting with fluorinated alkyl, aryl or vinyl halides using Zn or Cd catalyst via Barbier-type reaction. The preparation of +a-hydroxyvinylstannanes, and indol-2-yltributylstannane.The conversion of organozirconiums to organostannanes.

Specifications

CAS 1461-22-9
Density 1.2 g/mL at 25°C
Boiling Point 171°C to 173°C (25 mmHg, lit.)
Molecular Formula C12H27ClSn
Refractive Index n20/D 1.492
Linear Formula [CH3(CH2)3]3SnCl
MDL Number MFCD00000521
Quantity 500 g
Synonym TBTC; Tributylchlorotin
Molecular Weight (g/mol) 325.51 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.