Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyltin chloride, MilliporeSigma™ Supelco™

Catalog No. 111019653 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9653 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9653 Supplier MilliporeSigma Supelco Supplier No. 442869

Tributyltin chloride is a salt derivative of tributyltin which is an organotin compound.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 1461-22-9
Vapor Pressure <0.01 mmHg (20°C)
Density 1.2 g/mL (at 25°C)
Flash Point 113°C
Physical Form Neat
Linear Formula [CH3(CH2)3]3 SnCl
MDL Number MFCD00000521
Boiling Point 171°C to 173°C (25 mmHg)
Synonym TBTC; Tributylchlorotin
Type Organic Pollutant Standard
Quantity 500 mg
Formula Weight 325.51
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.