Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tributyltin Hydride

Catalog No. 234788500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-478-8500G 500 g
23-478-810G 10 g
23-478-850G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 23-478-8500G Supplier Sigma Aldrich Supplier No. 234788500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Radical promoted intramolecular cyclization leading to isoxazolo-benzaulene ring system. Vinyl tin fragment used in a synthesis of (+)-panepophenanthrin via biomimetic Diels-Alder dimerization.

Specifications

CAS 688-73-3
Density 1.082 g/mL at 25°C
Boiling Point 80°C (0.4 mmHg, lit.)
Molecular Formula C12H28Sn
Refractive Index n20/D 1.473
Linear Formula [CH3(CH2)3]3SnH
MDL Number MFCD00009416
Quantity 500 g
Molecular Weight (g/mol) 291.06 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.