Learn More
Apexbio Technology LLC Tributyrin 60-01-5 5ml
SDP
Catalog No. 5000097172
Encompass_Preferred
Tributyrin (CAS 60-01-5) is a triglyceride ester of butyric acid that functions as a prodrug delivering butyrate upon hydrolysis Butyrate is known to act as a histone deacetylase (HDAC) inhibitor thereby modulating gene expression and promoting cellular differentiation and apoptosis Tributyrin has been investigated in preclinical and clinical studies for its potential to inhibit tumor growth including applications in prostate cancer and adult solid tumors Due to its ability to release bioactive butyrate in vivo it serves as a valuable tool compound for studying epigenetic regulation and cancer therapeutics
Catalog No. 50-000-97172 Supplier Apexbio Technology LLC Supplier No. A46385
Add to Cart
Add to Cart
Tributyrin (CAS 60-01-5) is a triglyceride ester of butyric acid that functions as a prodrug delivering butyrate upon hydrolysis Butyrate is known to act as a histone deacetylase (HDAC) inhibitor thereby modulating gene expression and promoting cellular differentiation and apoptosis Tributyrin has been investigated in preclinical and clinical studies for its potential to inhibit tumor growth including applications in prostate cancer and adult solid tumors Due to its ability to release bioactive butyrate in vivo it serves as a valuable tool compound for studying epigenetic regulation and cancer therapeutics