Learn More
Sigma Aldrich Fine Chemicals Biosciences Tributyrin Agar, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 91015100G
Encompass
For the detection and enumeration of lipolytic microorganisms in food and other material (staphylococci, pseudomonads, clostridiae and marine flavobacteria).
Catalog No. NC0936066