Learn More
Medchemexpress LLC Tributyrin (glyceryl tributyrate) | 60-01-5 | 98.91% | 302.36 | 5 ML
SDP
Catalog No. 5000455037
Encompass_Preferred
Tributyrin (Glyceryl tributyrate) is a neutral short-chain fatty acid triglyceride that serves as a stable and rapidly absorbed proagent of Butyric Acid. It diffuses through biological membranes and is metabolized by intracellular lipases, which releases effective butyrate directly into the cell in vivo. Tributyrin has potent antiproliferative, proapoptotic, and differentiation-inducing effects.
  • Enhances antiproliferative effects of dihydroxycholecalciferol in human colon cancer cells.
  • Induces differentiation, growth arrest, and apoptosis in androgen-sensitive and androgen-resistant human prostate cancer cell lines.
  • Attenuates metabolic and inflammatory changes associated with obesity through a GPR109A-dependent mechanism.
  • Alleviates gut microbiota dysbiosis to repair intestinal damage in antibiotic-treated mice.
Catalog No. 50-004-55037 Supplier Medchemexpress LLC Supplier No. HYW0114045ML
Tributyrin (Glyceryl tributyrate) is a neutral short-chain fatty acid triglyceride that serves as a stable and rapidly absorbed proagent of Butyric Acid. It diffuses through biological membranes and is metabolized by intracellular lipases, which releases effective butyrate directly into the cell in vivo. Tributyrin has potent antiproliferative, proapoptotic, and differentiation-inducing effects.
  • Enhances antiproliferative effects of dihydroxycholecalciferol in human colon cancer cells.
  • Induces differentiation, growth arrest, and apoptosis in androgen-sensitive and androgen-resistant human prostate cancer cell lines.
  • Attenuates metabolic and inflammatory changes associated with obesity through a GPR109A-dependent mechanism.
  • Alleviates gut microbiota dysbiosis to repair intestinal damage in antibiotic-treated mice.