Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyrin, MP Biomedicals™

Catalog No. MP210311191 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210311191 1 L
MP210311190 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP210311191 Supplier MP Biomedicals, Inc Supplier No. 0210311191
Add to Cart
Add to Cart

Used as a deliverable source of natural butyrate. Tributyrin, MP Biomedicals is glycerol substituted with 3 butyrl esters. It induces differentiation of malignant cells, followed by arrest of cell growth and apoptosis. It potently inhibits growth of and i

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-01-5
Molecular Formula C15H26O6
Molecular Weight (g/mol) 302.367
InChI Key UYXTWWCETRIEDR-UHFFFAOYSA-N
Synonym tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin
PubChem CID 6050
ChEBI CHEBI:35020
IUPAC Name 2,3-di(butanoyloxy)propyl butanoate
SMILES CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC

Specifications

Melting Point -75°C (literature)
Density 1.032g/mL (Lit.)
Boiling Point 287°C to 288°C (literature)
Refractive Index n20/D 1.435 (Lit.)
Quantity 1 L
Solubility Information Soluble in alcohol and ether; insoluble in water.
Formula Weight 302.36
Percent Purity ≥96%
Physical Form Liquid
Recommended Storage Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.