Learn More
Syndel USA Syncaine 100g
SDP
Catalog No. NC0872873
Encompass
Syndels Syncaine (also known as MS-222, TMS, tricaine methanesulfonate) is a fish anesthetic used for the temporary immobilization of fish, amphibians, and other aquatic cold-blooded animals (poikilotherms) around the world. Syncaine has long been recognized as a valuable tool for the proper handling of these animals during manual spawning (fish stripping), weighing, measuring, marking, surgical operations, transport, photography, and research. Syncaine is a fully approved drug in the USA, Canada and the United Kingdom.
Catalog No. NC0872873 Supplier Syndel USA Supplier No. SYNCMGRUS02
Add to Cart
Add to Cart
Syndels Syncaine (also known as MS-222, TMS, tricaine methanesulfonate) is a fish anesthetic used for the temporary immobilization of fish, amphibians, and other aquatic cold-blooded animals (poikilotherms) around the world. Syncaine has long been recognized as a valuable tool for the proper handling of these animals during manual spawning (fish stripping), weighing, measuring, marking, surgical operations, transport, photography, and research. Syncaine is a fully approved drug in the USA, Canada and the United Kingdom.