Learn More
PENTAIR AQUATIC ECO SYSTEMS INC TRICAINE-S (MS222) 100G
Catalog No. NC0342409
Encompass
Manufactured by Syndel USA. Tricaine-S Topical Anesthetics (MS-222) is a great sedative for transporting, sorting and grading fish. This is an FDA-approved, ANADA #200-226, high-grade brand of MS-222 (tricaine methanesulfonate), which is an anesthetic/tranquilizer of fish and other cold-blooded aquatic organisms. When used properly, it induces a temporary state of immobilization that is rapidly reversed when the animal is placed back in clean water. Commonly used when handling fish during manual spawning (stripping), tagging, measuring, weighing and surgical operations. Tricaine-S is a USFDA registered drug of Western Chemical Inc. a subsidiary of Aquatic Life Sciences, Inc.
Catalog No. NC0342409 Supplier PENTAIR AQUATIC ECO SYSTEMS INC Supplier No. TRS1
Manufactured by Syndel USA. Tricaine-S Topical Anesthetics (MS-222) is a great sedative for transporting, sorting and grading fish. This is an FDA-approved, ANADA #200-226, high-grade brand of MS-222 (tricaine methanesulfonate), which is an anesthetic/tranquilizer of fish and other cold-blooded aquatic organisms. When used properly, it induces a temporary state of immobilization that is rapidly reversed when the animal is placed back in clean water. Commonly used when handling fish during manual spawning (stripping), tagging, measuring, weighing and surgical operations. Tricaine-S is a USFDA registered drug of Western Chemical Inc. a subsidiary of Aquatic Life Sciences, Inc.