Learn More
PENTAIR AQUATIC ECO SYSTEMS INC Tricaine-S Topical Anesthetics, 10 Gram
Supplier:  PENTAIR AQUATIC ECO SYSTEMS INC TRS5
Encompass
Manufactured by Syndel USA. Tricaine-S Topical anesthetics (MS-222). This is an FDa-approved, aNaDa #200-226, high-grade brand of MS-222 (tricaine methanesulfonate), which is an anesthetic/tranquilizer of fish and other cold-blooded aquatic organisms. When used properly, it induces a temporary state of immobilization that is rapidly reversed when the animal is placed back in clean water. Commonly used when handling fish during manual spawning (stripping), tagging, measuring, weighing and surgical operations. Tricaine-S is a USFDa registered drug of Western Chemical Inc. a subsidiary of aquatic Life Sciences, Inc
Catalog No. NC0589195