Learn More
TARGETMOL CHEMICALS INC TRICAPRILIN 1000MG
SDP
Catalog No. 502576696
Encompass_Preferred
Also available in 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tricaprilin (Glycerol Tri-n-octanoate) is used in study for patients with mild to moderate Alzheimer's disease. Tricaprilin also has a role as an anticonvulsant and a plant metabolite. purity: 98%
Catalog No. 50-257-6696 Supplier TARGETMOL CHEMICALS INC Supplier No. T52661G
Also available in 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tricaprilin (Glycerol Tri-n-octanoate) is used in study for patients with mild to moderate Alzheimer's disease. Tricaprilin also has a role as an anticonvulsant and a plant metabolite. purity: 98%