Learn More
Medchemexpress LLC Tricaproin 1mL
SDP
Catalog No. 5000204947
Encompass_Preferred
TricaproinIt is an organic compound belonging to the class of triacylglycerols It consists of three molecules of caproic acid esterified to the glycerin backbone with a mild fatty smell TricaproinIt has a variety of applications in the cosmetic and personal care industries especially as an emollient and conditioning agent in skin and hair care products Additionally it is used as a lubricant and surfactant in industrial settings
Catalog No. 50-002-04947 Supplier Medchemexpress LLC Supplier No. HYW1275341ML
TricaproinIt is an organic compound belonging to the class of triacylglycerols It consists of three molecules of caproic acid esterified to the glycerin backbone with a mild fatty smell TricaproinIt has a variety of applications in the cosmetic and personal care industries especially as an emollient and conditioning agent in skin and hair care products Additionally it is used as a lubricant and surfactant in industrial settings