Learn More
MOLECULAR DEPOT LLC Tricaprylin Triglyceride 200mg
Catalog No. 5000013483
Encompass_Preferred
Tricaprylin Triglyceride (200 mg) is a triester of glycerin and octanoic acid with a molecular weight of 470 69 g/mol It is supplied as a neat liquid For in vitro research use only Not for human or animal use
Catalog No. 50-000-13483 Supplier MOLECULAR DEPOT LLC Supplier No. B2010629
Tricaprylin Triglyceride (200 mg) is a triester of glycerin and octanoic acid with a molecular weight of 470 69 g/mol It is supplied as a neat liquid For in vitro research use only Not for human or animal use