Learn More
Medchemexpress LLC Tricarballylic acid | 99-14-9 | MFCD00002723 | 98.0% | 176.12 g/mol | C6H8O6 | 5 G
SDP
Catalog No. 5000300738
Encompass_Preferred
Tricarballylic acid is a tricarboxylic acid used in biochemical research as an aconitate hydratase inhibitor and analytical standard. It is supplied in solid and solution formats for laboratory use and is employed in enzyme inhibition studies, metabolic research, and analytical calibration.
  • Inhibits aconitate hydratase for enzymatic studies.
  • Available as solid or solution standards for analytical use.
  • High purity suitable for research applications.
  • Well-characterized molecular properties for reproducible results.
  • Compatible with common organic solvents for stock preparation.
Catalog No. 50-003-00738 Supplier Medchemexpress LLC Supplier No. HYW0202155G
Add to Cart
Add to Cart
Tricarballylic acid is a tricarboxylic acid used in biochemical research as an aconitate hydratase inhibitor and analytical standard. It is supplied in solid and solution formats for laboratory use and is employed in enzyme inhibition studies, metabolic research, and analytical calibration.
  • Inhibits aconitate hydratase for enzymatic studies.
  • Available as solid or solution standards for analytical use.
  • High purity suitable for research applications.
  • Well-characterized molecular properties for reproducible results.
  • Compatible with common organic solvents for stock preparation.