Learn More
Medchemexpress LLC Tricarbonylchloro(glycinato)ruthenium | 475473-26-8 | 97.5% | 294.61 g/mol | C5H4ClNO5Ru | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10058110MM 1ML
Encompass_Preferred
CORM-3 is a carbon monoxide-releasing molecule used in research to deliver controlled CO to biological systems. It modulates inflammatory signaling and oxidative stress pathways, making it a tool for studying CO-mediated cytoprotective and anti-inflammatory effects.
  • Releases carbon monoxide for biological studies.
  • Attenuates NF-κB p65 nuclear translocation.
  • Reduces generation of reactive oxygen species.
  • Enhances intracellular glutathione and superoxide dismutase levels.
  • Reduces NLRP3 inflammasome activation.
  • Available as a 10 mM solution in DMSO (1 mL) and as solid quantities for synthesis or formulation.
Catalog No. 50-002-12345