Learn More
Medchemexpress LLC Tricarbonylchloro(glycinato)ruthenium (II) | 475473-26-8 | 97.5% | 294.61 | C5H4ClNO5Ru | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10058150MG
Encompass_Preferred
CORM-3 is a water-soluble carbon monoxide (CO) releasing molecule used in research to study the biological effects of CO, including anti-inflammatory and cardioprotective responses. It is suitable for controlled CO delivery in cellular and biochemical assays and is supplied in solid or solution formats for laboratory use.
  • Water-soluble CO-releasing molecule for biological studies.
  • Suitable for in vitro studies of NF-κB signaling and reactive oxygen species.
  • Available as solid powder or pre-made solution for flexible workflows.
  • High purity supports reproducible experimental results.
  • Compatible with aqueous buffers and cellular media for rapid CO release.
Catalog No. 50-002-12992