Learn More
Medchemexpress LLC Tricarbonylchloro(glycinato)ruthenium (II) | 475473-26-8 | MFCD07364048 | 97.5% | 294.61 g·mol⁻¹ | C5H4ClNO5Ru | 5 MG
SDP
Catalog No. 5000213342
Encompass_Preferred
CORM-3 (tricarbonylchloro(glycinato)ruthenium (II)) is a water-soluble carbon monoxide-releasing molecule supplied as a powdered research reagent. It is used to deliver controlled doses of CO in biochemical and cellular studies, enabling investigation of CO-mediated signaling, redox biology, and cytoprotective effects under laboratory conditions.
  • Releases carbon monoxide under physiological conditions.
  • Water-soluble ruthenium carbonyl complex for aqueous studies.
  • Suitable for biochemical, cellular, and physiological research models.
  • Provided as a stable powder for frozen or cold storage.
  • High reported purity for reproducible experimental results.
  • Molecular weight ~294.6 g·mol⁻¹; formula C5H4ClNO5Ru.
Catalog No. 50-002-13342 Supplier Medchemexpress LLC Supplier No. HY1005815MG
CORM-3 (tricarbonylchloro(glycinato)ruthenium (II)) is a water-soluble carbon monoxide-releasing molecule supplied as a powdered research reagent. It is used to deliver controlled doses of CO in biochemical and cellular studies, enabling investigation of CO-mediated signaling, redox biology, and cytoprotective effects under laboratory conditions.
  • Releases carbon monoxide under physiological conditions.
  • Water-soluble ruthenium carbonyl complex for aqueous studies.
  • Suitable for biochemical, cellular, and physiological research models.
  • Provided as a stable powder for frozen or cold storage.
  • High reported purity for reproducible experimental results.
  • Molecular weight ~294.6 g·mol⁻¹; formula C5H4ClNO5Ru.