Learn More
Medchemexpress LLC HY-W033577 25mg Medchemexpress, Tricarbonyldichlororuthenium(II) dimer CAS:22594-69-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW03357725MG
Encompass_Preferred
Medchemexpress, HY-W033577 25mg Tricarbonyldichlororuthenium(II) dimer CAS:22594-69-0 Tricarbonyldichlororuthenium(II) dimer is a pharmacological donor of CO releasing. CO releases from Tricarbonyldichlororuthenium(II) dimer prevents gastric mucosal oxidative damage induced by ischemia/reperfusion (I/R) improving gastric blood flow (GBF), decreasing DNA oxidation and inflammatory response on systemic level. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3450