Learn More
Apexbio Technology LLC Trichlormethiazide 133-67-5 1g
SDP
Catalog No. 5000092958
Encompass_Preferred
Trichlormethiazide is a small-molecule inhibitor targeting the sodium-chloride cotransporter (NCC) It is designed to inhibit NCC in the distal convoluted tubules thereby decreasing sodium and chloride reabsorption and modulating fluid and electrolyte balance Trichlormethiazide exerts its biological activity primarily through inhibition of sodium and chloride reabsorption Based on these pharmacological properties Trichlormethiazide holds research potential in studies of electrolyte and fluid regulation hypertension-related pathophysiology and renal function as well as the investigation of renal tubular sodium handling and screening interventions affecting nephron sodium transport processes in biological models
Catalog No. 50-000-92958 Supplier Apexbio Technology LLC Supplier No. B20561000
Trichlormethiazide is a small-molecule inhibitor targeting the sodium-chloride cotransporter (NCC) It is designed to inhibit NCC in the distal convoluted tubules thereby decreasing sodium and chloride reabsorption and modulating fluid and electrolyte balance Trichlormethiazide exerts its biological activity primarily through inhibition of sodium and chloride reabsorption Based on these pharmacological properties Trichlormethiazide holds research potential in studies of electrolyte and fluid regulation hypertension-related pathophysiology and renal function as well as the investigation of renal tubular sodium handling and screening interventions affecting nephron sodium transport processes in biological models