Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Trichlormethiazide USP
SDP

Catalog No. 14185620 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-620 25 g
14-185-430 500 g
14-185-284 100 g
14-185-287 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-185-620 Supplier Medisca Supplier No. 026804
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trichlormethiazide USP is a white or practically white, crystalline powder. It is odorless to having a slight characteristic odor.

Trichlormethiazide USP is a white or practically white, crystalline powder. It is odorless to having a slight characteristic odor.

Specifications

Chemical Name or Material Trichlormethiazide USP
Packaging Jar, 50mL, White, HDPE, Round, 38-400, TE Pano Neck
CAS 133-67-5
Type Drug Standard
Molecular Weight (g/mol) 380.659
Quantity 25 g
Percent Purity 98 to 102 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.