Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloro(1H,1H,2H,2H-heptadecafluorodecyl)silane 96.0+%, TCI America™
SDP

Catalog No. T270525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T270525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. T270525G Supplier TCI America Supplier No. T270525G

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 78560-44-8
Color Colorless
Boiling Point 118°C
Molecular Formula C10H4Cl3F17Si
Quantity 25 g
MDL Number MFCD00042344
UN Number 2987
Synonym 1h,1h,2h,2h-perfluorodecyltrichlorosilane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl, 1h,1h,2h,2h-perfluorotrichlorosilane, trichloro 1h,1h,2h,2h-perfluorodecyl silane, trichloro 1h,1h,2h,2h-heptadecafluorodecyl silane, 1-trichlorosilyl-1h,1h,2h,2h-heptadecafluorodecane, 1h,1h,2h,2h-perfluorodecyltrichlorosilane 1g, heptadecafluoro-1,1,2,2-tetrahydrodecyl trichlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl-1-trichlorosilane
InChI Key VIFIHLXNOOCGLJ-UHFFFAOYSA-N
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Si](Cl)(Cl)Cl
IUPAC Name trichloro(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
Molecular Weight (g/mol) 581.55
PubChem CID 123577
Formula Weight 581.55
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Trichloro(1H,1H,2H,2H-heptadecafluorodecyl)silane
Show More Show Less
EINECSNumber (2)-2046
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.