Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trichloro(3,3,3-Trifluoropropyl)Silane

Catalog No. 45280725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-280-725ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 45-280-725ML Supplier Sigma Aldrich Supplier No. 45280725ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trichloro(3,3,3-trifluoropropyl)silane can be used as a precursor for the preparation of: Fluorinated polyhedral oligomeric silsesquioxane (F-POSS) by base-catalyzed condensation 3,3,3-Trifluoropropyltrimethoxysilane by alcoholysis reaction with methanol.It can also be used as a silanizing agent to coat the inner surface of pipettes in order to investigate the electrical charging of droplets discharged from the micropipette.

Specifications

CAS 73852-19-4
Melting Point 266°C to 270°C
Molecular Formula C5H12N2O -+ HCl
Refractive Index n20/D 1.386
Linear Formula (CH3)2CHCH(NH2)CONH2 -+ HCl
MDL Number MFCD00039085
Quantity 5 g
Synonym 3,3,3-Trifluoropropyl-trichlorosilane
Molecular Weight (g/mol) 152.62 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.