Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloro(3-phenylpropyl)silane 97.0+%, TCI America™
SDP

Catalog No. T28535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T28535G Supplier TCI America Supplier No. T28535G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13617-40-8
Molecular Formula C9H11Cl3Si
Molecular Weight (g/mol) 253.622
MDL Number MFCD09953886
InChI Key QJOHXSVBLYVERP-UHFFFAOYSA-N
Synonym (3-Phenylpropyl)trichlorosilane
PubChem CID 12441988
IUPAC Name trichloro(3-phenylpropyl)silane
SMILES C1=CC=C(C=C1)CCC[Si](Cl)(Cl)Cl

Specifications

Color Colorless
Boiling Point 126°C
Quantity 5 g
UN Number 2987
Formula Weight 253.62
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trichloro(3-phenylpropyl)silane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.