Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trichloro(octadecyl)silane

Catalog No. 104817500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-481-7500G 500 g
10-481-7100G 100 g
10-481-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-481-7500G Supplier Sigma Aldrich Supplier No. 104817500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trichloro(octadecyl)silane (OTS) is an alkoxy silane with a large alkyl and polar head group, mainly used as a self-assembled monolayer (SAM) on a variety of nanoparticles and metal oxides substrates. It provides an amphiphilic character that can be used in molecular and opto-electronics applications.OTS can be used to silanize poly(vinylalcohol) (PVA)/nanosilica (SiO2) nanofibers that can provide a hydrophobic coating on the wooden surfaces by electrospinning. It can also be used as a SAM on the surface of silica (SiO2) gate dielectric for the development of organic field effect transistor (OFETs). It may also be used in a variety of applications like improving the electrochemical performance for dye sensitized solar cells and for the development of metal insulated field effect transistors.

Specifications

CAS 112-04-9
Density 0.984 g/mL (at 25°C)
Boiling Point 223°C
Molecular Formula C18H37Cl3Si
Refractive Index n20/D 1.459 (literature)
Linear Formula CH3(CH2)17 SiCl3
Quantity 500 g
MDL Number MFCD00000484
Synonym Octadecyltrichlorosilane
Molecular Weight (g/mol) 387.93
Percent Purity ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.