Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trichloro(octyl)silane

Catalog No. 235725100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-572-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-572-5100G Supplier Sigma Aldrich Supplier No. 235725100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trichloro(octyl)silane (OTS) is a linear alkylsilane which is used as a modifying agent which creates a low cost hydrophobic surface with high thermo-mechanical stability. OTS contains hydrolysable groups which form a covalent bond with the hydroxyl groups present on the surface.OTS functionalized magnesium oxide (MnO2)/aluminium oxide (Al2O3) surface can be used as a catalyst in the decomposition of ozone (O3) and nitrogen dioxide (NO2). OTS can also be used for the surface modification of silicon oxide (SiO2), used in the fabrication of pentacene organic field effect transistors (OFETs). OTS functionalized wafers can be coated on enhanced green fluorescent protein (GFP) and anchor peptides based films to determine the thickness of the films.

Specifications

CAS 5283-66-9
Density 1.07 g/mL (at 25°C)
Boiling Point 233°C (731 mmHg)
Molecular Formula C8H17Cl3Si
Linear Formula CH3(CH2)7 SiCl3
Refractive Index n20/D 1.447 (literature)
Quantity 100 g
MDL Number MFCD00000488
Synonym Octyltrichlorosilane
Molecular Weight (g/mol) 247.67
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.