Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trichloro(phenethyl)silane

Catalog No. 4200341G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-003-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-003-41G Supplier Sigma Aldrich Supplier No. 4200341G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trichloro(phenethyl)silane (PTS) is a silane based capping agent that can be used to form a self-assembled monolayer (SAM) on a variety of surfaces. It facilitates the surface treatment and modification for effective detection of surface atoms.PTS forms a SAM on a wide range of substrates such as aluminum oxide and silica, which can be further coated on organic field effect transistors (OFETs), thin film transistors and other biosensor based devices for biomedical applications.

Specifications

CAS 940-41-0
Density 1.234 g/mL (at 25°C)
Boiling Point 93°C to 96°C (3 mmHg)
Molecular Formula C8H9Cl3Si
Refractive Index n20/D 1.513 (literature)
Linear Formula C6H5CH2CH2SiCl3
Quantity 1 g
MDL Number MFCD00039290
Synonym (2-Phenylethyl)trichlorosilane; Phenethyltrichlorosilane
Molecular Weight (g/mol) 239.6
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.