Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trichloro(Phenyl)Silane

Catalog No. 4401081L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-010-81L 1 L
44-010-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 44-010-81L Supplier Sigma Aldrich Supplier No. 4401081L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trifluoro(phenyl)silane can react with aliphatic alcohols to yield benzene and alkoxyfluoro and tetraalkoxysilanes. Hydrolytic condensationof this silane in its vapor phase results in the formation of 1,1,3,3-Tetrachloro-1,3-diphenyldisiloxane.

Specifications

CAS 98-13-5
Density 1.321 g/mL at 25°C
Boiling Point 201°C
Molecular Formula C6H5Cl3Si
Linear Formula C6H5SiCl3
Refractive Index n20/D 1.523
MDL Number MFCD00000479
Quantity 1 L
Synonym Phenyltrichlorosilane
Molecular Weight (g/mol) 211.55 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.