Learn More
Crescent Chemical Co Inc TRICHLOROACETIC ACID 20% SOL
SDP
Catalog No. 50248211
Encompass_Preferred
Trichloroacetic acid, 20 % solution Aqueous solution. TCA (cat. no. 36910): 200 g/L. Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 36913.02
Catalog No. 50-248-211 Supplier Crescent Chemical Co Inc Supplier No. SE36913.02
Trichloroacetic acid, 20 % solution Aqueous solution. TCA (cat. no. 36910): 200 g/L. Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 36913.02