Learn More
Sigma Aldrich Fine Chemicals Biosciences Trichloroacetic acid BioUl100G
Catalog No. 502899411
Encompass
Trichloroacetic acid (TCA) is derived from Trichloroethylene (TCE) metabolism. It is used as an acid decalcifying agent. TCA is used as a fixative for nuclear staining and protein precipitation.
Catalog No. 50-289-9411 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 91228100G
Trichloroacetic acid (TCA) is derived from Trichloroethylene (TCE) metabolism. It is used as an acid decalcifying agent. TCA is used as a fixative for nuclear staining and protein precipitation.