Learn More
Sigma Aldrich Fine Chemicals Biosciences Trichloroacetic acid BioUl500G
Catalog No. 502898613
Encompass
Trichloroacetic acid (TCA) is derived from Trichloroethylene (TCE) metabolism. It is used as an acid decalcifying agent. TCA is used as a fixative for nuclear staining and protein precipitation.
Catalog No. 50-289-8613 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 91228500G
Trichloroacetic acid (TCA) is derived from Trichloroethylene (TCE) metabolism. It is used as an acid decalcifying agent. TCA is used as a fixative for nuclear staining and protein precipitation.