Learn More
Sigma Aldrich Fine Chemicals Biosciences Trichloroacetic acid suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), >=99% | 76-03-9 | MFCD00004177 | 250G
Catalog No. 501847370
Encompass
Trichloroacetic acid suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), >=99% | Purity: >=99% | Mol Wt: 163.39 | 76-03-9 | MFCD00004177 | 250G
Catalog No. 50-184-7370 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T8657250G
Trichloroacetic acid suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), >=99% | Purity: >=99% | Mol Wt: 163.39 | 76-03-9 | MFCD00004177 | 250G