Learn More
Sigma Aldrich Fine Chemicals Biosciences Trichloroacetic acid suita500G
Catalog No. 502899907
Encompass
Trichloroacetic acid (TCA) is produced during the chlorination of water comprising natural organic material. It is a novel reagent used in clinical laboratories for deproteinization of body fluids. In addition TCA is also used in the treatment of genital warts and acne scars.
Catalog No. 50-289-9907 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T8657500G
Trichloroacetic acid (TCA) is produced during the chlorination of water comprising natural organic material. It is a novel reagent used in clinical laboratories for deproteinization of body fluids. In addition TCA is also used in the treatment of genital warts and acne scars.