Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trichloroacetic anhydride

Catalog No. 36771025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-771-025G 25 g
36-771-0100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-771-025G Supplier Sigma Aldrich Supplier No. 36771025G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trichloroacetic anhydride is reported to be an efficient derivatization reagent, since it produces stable derivatives having both a higher mass fragmentation pattern and lesser volatility than derivatives of trifluoroacetic anhydride (tfaa) or heptafluorobutyric anhydride (hfba). the interaction of thymidine 5α-phosphate with trichloroacetic anhydride in dimethylformamide and acetonitrile in the presence of tertiary amines hs been studied.trichloroacetic anhydride may be used as derivatization reagent for the following studies:
  • Simultaneous analysis of amphetamine, methamphetamine and 3,4-methylenedioxymethamphetamine (mdma or ecstasy) in urine samples by solid-phase extraction, derivatization and gas chromatography/mass spectrometry
  • Analysis of halogenated amphetamines in brain tissue from rats by gas chromatography with electron-capture detection (GC-ecd)
  • Quantitative determination of the metabolites of L-alpha-acetylmethadol (laam), such as noracetylmethadol, dinoracetylmethadol, methadol and normethadol by electron capture gas-liquid chromatography

Specifications

CAS 4124-31-6
Density 1.69 g/mL (at 25°C (literature))
Boiling Point 139°C to 141°C (60 mmHg)
Molecular Formula C4Cl6O3
Refractive Index n20/D 1.484 (literature)
Linear Formula (Cl3CCO)2 O
MDL Number MFCD00000793
Quantity 25 g
Molecular Weight (g/mol) 308.76
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.