Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trichloroacetonitrile

Catalog No. T53805500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T53805500G 500 g
T53805100G 100 g
T538055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. T53805500G Supplier Sigma Aldrich Supplier No. T53805500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Employed in a synthesis of trichloroacetimidates by DBU catalyzed addition of allylic alcohols and the subsequent study of the MOM catalyzed aza-Claisen rearrangement. Also used to prepare bistrichloroacetimidates from diols leading to dihyrooxazines via acid catalyzed cyclization.

Specifications

CAS 545-06-2
Density 1.44 g/mL at 25°C
Boiling Point 83°C to 84°C
Molecular Formula C2Cl3N
Refractive Index n20/D 1.441
Linear Formula Cl3CCN
MDL Number MFCD00001842
Quantity 500 g
Molecular Weight (g/mol) 144.39 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.