Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trichloroacetyl isocyanate

Catalog No. 9109525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-095-25ML 25 mL
91-095-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 91-095-25ML Supplier Sigma Aldrich Supplier No. 9109525ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Derivatizing reagent for the structural assignment to hydroxy compounds by NMR; Used in negative ion chemical ionization mass spectrometry

Specifications

CAS 3019-71-4
Density 1.581 g/mL (at 25°C (literature))
Boiling Point 80°C to 85°C (20 mmHg)
Molecular Formula C3Cl3NO2
Refractive Index n20/D 1.480 (literature); n20/D 1.480
Linear Formula Cl3CCONCO
MDL Number MFCD00002033
Quantity 25 mL
Molecular Weight (g/mol) 188.4
Percent Purity ≥97.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.