Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichlorocyclopentylsilane 95.0+%, TCI America™
SDP

Catalog No. T257625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T257625G 25 g
T25765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T257625G Supplier TCI America Supplier No. T257625G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14579-03-4
Molecular Formula C5H9Cl3Si
Molecular Weight (g/mol) 203.562
MDL Number MFCD00236508
InChI Key FCMZRNUHEXJWGB-UHFFFAOYSA-N
Synonym cyclopentyltrichlorosilane, trichloro cyclopentyl silane, silane, trichlorocyclopentyl, cyclopentane, trichlorosilyl, trichlorosilylcyclopentane, cyclopentyl trichlorosilane, acmc-1c6jw, trichlorocyclopentylsilane
PubChem CID 84533
IUPAC Name trichloro(cyclopentyl)silane
SMILES C1CCC(C1)[Si](Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 178°C
Quantity 25 g
UN Number 2987
Formula Weight 203.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trichlorocyclopentylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.