Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroethylene, MilliporeSigma™ Supelco™

Catalog No. 111019990 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9990 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-9990 Supplier MilliporeSigma Supelco Supplier No. 462675MLR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Trichloroethylene is a volatile and chlorinated organic compound, which finds applications as a solvent and greasing agent.

Cleaning Products, Cosmetics, Environmental, Food and Beverages, Personal Care and Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 770°F
CAS 79-01-6
Vapor Pressure 61 mmHg (20°C)
Vapor Density 4.5 (vs. air)
Density 1.463 g/mL (at 25°C)
Physical Form Neat
Linear Formula ClCH=CCl2
MDL Number MFCD00000838
Melting Point -84.8°C
Boiling Point 86.7°C
Synonym TCE; Trichloroethene
Type Analytical Standard
Quantity 5 mL
Formula Weight 131.39
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.