Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroethylene, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111017853 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7853 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-7853 Supplier MilliporeSigma Supelco Supplier No. PHR15623X1.2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. Trichloroethylene (TCE) is a chlorine containing organic compound, widely employed as an industrial solvent. It is formed as a major intermediate during the biodegradation of tetrachloroethylene (PCE) in a small anaerobic continuous-flow fixed film column.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 79-01-6
Vapor Pressure 61 mmHg (20°C)
Vapor Density 4.5 (vs. air)
Density 1.463 g/mL (at 25°C)
Linear Formula ClCH=CCl2
MDL Number MFCD00000838
Melting Point -84.8°C
Boiling Point 86.7°C
Synonym Trichloroethylene; TCE; Trichloroethene
Quantity 3 x 1.2 mL
Formula Weight 131.39
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.