Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trichlorosilane

Catalog No. 1755525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-555-25G 5 g
17-555-2500G 500 g
17-555-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 17-555-25G Supplier Sigma Aldrich Supplier No. 1755525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trichlorosilane is generally used for the asymmetric hydrosilylation of olefins in the presence of palladium catalysts coordinated with chiral monodentate phosphorus ligands to generate chiral organosilanes.Trichlorosilane has been used to synthesize 11-dicyclohexylphosphino-12-phenyl-9,10-dihydro-9,10-ethenoanthracene (H-KITPHOS) via reduction of 11-dicyclohexylphosphinoyl-12-phenyl-9,10-dihydro-9,10-ethenoanthracene.Other possible applications:Asymmetric reduction of N-aryl ketimines in the presence of a novelL-valine-derived catalyst to form secondary amines.Hydrosilylation of imidazolinones to form chiral imidazolidinones in the presence of a 2,2'sa-bispyrrolidine based Lewis base organocatalyst.Trichlorosilane activated with chiral N-formylprolinederivatives is an effective reagent for the reduction of imines to formenantiomerically enriched amines.Trichlorosilane reacts with dimethylformamide to form hypervalent hydridosilicates, which can reduce aldehydes to alcohols, imines to amines, and also for the reductive amination of aldehydes.

Specifications

CAS 10025-78-2
Density 1.342 g/mL at 25°C
Boiling Point 32°C to 34°C
Molecular Formula HCl3Si
Linear Formula SiHCl3
MDL Number MFCD00011518
Quantity 5 g
Molecular Weight (g/mol) 135.45 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.