Learn More
Trichlorosilane, 98%
Description
Trichlorosilane is used to prepare silicon for semiconductors. It is an active ingredient used in the preparation of polysilicons and silicone resins. It acts as coupling agents and silane adhesion promoter. It is a precursor to prepare organosilicon compounds through hydrosilylation reactions. Further, it is involved in the preparation of octadecyltrichlorosilane, perfluoroctyltrichlorosilane, and perfluordecyltrichlorosilane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 31°C to 32°C |
| Melting Point | -127°C |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 500 g |
| UN Number | UN1295 |
| Flash Point | -28°C (7°F) |
| Merck Index | 14,9646 |
| Solubility Information | Soluble in benzene,ether,heptane,chloroform and carbon tetrachloride. |
| Formula Weight | 135.45 |
| Show More |
Safety and Handling
| Hazard Category | H224-H250-H302-H314-H331-H335 |
| Hazard Statement | GHS H Statement H224-H250-H314-H302-H332; Extremely flammable liquid and vapour. Catches fire spontaneously if exposed to air. Causes severe skin burns and eye damage. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P210-P222-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: TRICHLOROSILANE |
| EINECSNumber | 233-042-5 |
| RTECSNumber | VV5950000 |
| TSCA | Yes |
| Recommended Storage | 2° to 8°C |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.