Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trichlorovinylsilane

Catalog No. 104876500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-487-6500G 500 g
10-487-6100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-487-6500G Supplier Sigma Aldrich Supplier No. 104876500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trichlorovinylsilane

Specifications

Boiling Point 90°C
Melting Point -95°C
CAS 75-94-5
Quantity 500 g
Molecular Formula C2H3Cl3Si
Linear Formula H2C=CHSiCl3
MDL Number MFCD00000482
Synonym (Trichlorosilyl)ethylene; Vinyltrichlorosilane
Molecular Weight (g/mol) 161.49
Percent Purity 97%
Refractive Index n20/D 1.436 (literature)
Density 1.27 g/mL (at 25°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.