Learn More
ATCC Trichomonas vaginalis; CDC 085
Supplier:  ATCC 50143
Encompass
Trichomonas vaginalis strain CDC 085 was isolated in Columbus, Ohio. This parasitic protozoan is resistant to metronidazole and can be used in antimicrobial resistance research, drug development, infectious disease research, and sexually transmitted disease research.
Catalog No. 50-238-4657
 Restricted Item