Learn More
ATCC Trichomonas vaginalis; G3
Supplier:  ATCC PRA98
Encompass
Trichomonas vaginalis strain G3 was isolated from a clinical specimen from Beckenham, United Kingdom. This parasitic protozoan has applications in infectious disease research and sexually transmitted disease research.
Catalog No. 50-238-2077