Learn More
ATCC Trichomonas vaginalis; G3; genomic DNA
Catalog No. 502384668
Encompass
Genomic DNA isolated from Trichomonas vaginalis strain G3 (ATCC PRA-98). This preparation of high molecular weight DNA can be used in PCR and other molecular biology applications.
Catalog No. 50-238-4668 Supplier ATCC Supplier No. PRA98D
Add to Cart
Add to Cart
Genomic DNA isolated from Trichomonas vaginalis strain G3 (ATCC PRA-98). This preparation of high molecular weight DNA can be used in PCR and other molecular biology applications.