Learn More
STEMCELL Technologies Trichostatin A, Size: 1 mg
SDP
Catalog No. NC1686746
Encompass
Inhibit histone deacetylase (HDAC)1 and HDAC6 with Trichostatin A (Furumai et al. Proc Natl Acad Sci USA, 2001; Yoshida et al. J Biol Chem, 1990). This epigenetic modifier increases the reprogramming efficiency of mouse embryonic fibroblasts to induced pluripotent stem (iPS) cells (Huangfu et al. Nat Biotechnol, 2008). It promotes differentiation of hepatocytes from human mesenchymal stem cells (Snykers et al. BMC Dev Biol, 2007) and prevents dedifferentiation of primary rat hepatocytes in culture, maintaining liver-specific cellular functions (Henkens et al. Toxicol Appl Pharmacol, 2007). Trichostatin A also resets epigenetic memory in mouse iPS cells, in combination with 5-Azacytidine (Kim et al. Nature, 2010). Trichostatin A is supplied as a crystalline solid with ≥ 98% purity; Molecular weight 302.4 g/mol; CAS number 58880-19-6. Visit STEMCELL.com for more information and related products.
Catalog No. NC1686746 Supplier STEMCELL Technologies Supplier No. 72282
Inhibit histone deacetylase (HDAC)1 and HDAC6 with Trichostatin A (Furumai et al. Proc Natl Acad Sci USA, 2001; Yoshida et al. J Biol Chem, 1990). This epigenetic modifier increases the reprogramming efficiency of mouse embryonic fibroblasts to induced pluripotent stem (iPS) cells (Huangfu et al. Nat Biotechnol, 2008). It promotes differentiation of hepatocytes from human mesenchymal stem cells (Snykers et al. BMC Dev Biol, 2007) and prevents dedifferentiation of primary rat hepatocytes in culture, maintaining liver-specific cellular functions (Henkens et al. Toxicol Appl Pharmacol, 2007). Trichostatin A also resets epigenetic memory in mouse iPS cells, in combination with 5-Azacytidine (Kim et al. Nature, 2010). Trichostatin A is supplied as a crystalline solid with ≥ 98% purity; Molecular weight 302.4 g/mol; CAS number 58880-19-6. Visit STEMCELL.com for more information and related products.