Learn More
Medchemexpress LLC HY-15144 2mg Medchemexpress, Trichostatin A CAS:58880-19-6 Purity:>98%
SDP
Catalog No. 502029082
Encompass_Preferred
Medchemexpress, HY-15144 2mg Trichostatin A CAS:58880-19-6 Trichostatin A (TSA) is a potent and specific inhibitor of HDAC class I/II, with an IC50 value of 1.8 nM for HDAC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9082 Supplier Medchemexpress LLC Supplier No. HY151442MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-15144 2mg Trichostatin A CAS:58880-19-6 Trichostatin A (TSA) is a potent and specific inhibitor of HDAC class I/II, with an IC50 value of 1.8 nM for HDAC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.