Learn More
Eurofins Discoverx Trichostatin A, 5 mg. (92-1246)
Supplier:  Eurofins Discoverx 921246
Encompass
Trichostatin A (TSA) is a naturally occurring antibiotic that inhibits the eukaryotic cell cycle during the growth stage. Potent and selective inhibitor of Class I and II histone deacetylases (HDAC, IC50=70 nM for HDAC1), inhibiting removal of acetyl groups from histones and resulting in hyperacetylation. Leads to chromatin relaxation and gene expression modulation. Provided as 5mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9767