Learn More
Medchemexpress LLC Trichostatin A | 58880-19-6 | C17H22N2O3 | 1 ML
SDP
Catalog No. 5000372184
Encompass_Preferred
Trichostatin A (TSA) is a potent and specific inhibitor of HDAC class I/II, exhibiting an IC50 value of 1.8 nM for HDAC. It is intended for research use only and not for patient applications.
  • Potent and specific inhibitor of HDAC class I/II
  • Inhibits proliferation in various carcinoma cell lines
  • Enhances osteogenic differentiation of ADSCs
  • Demonstrates in vivo antitumor activity
Catalog No. 50-003-72184 Supplier Medchemexpress LLC Supplier No. HY1514410MM/1ML
Trichostatin A (TSA) is a potent and specific inhibitor of HDAC class I/II, exhibiting an IC50 value of 1.8 nM for HDAC. It is intended for research use only and not for patient applications.
  • Potent and specific inhibitor of HDAC class I/II
  • Inhibits proliferation in various carcinoma cell lines
  • Enhances osteogenic differentiation of ADSCs
  • Demonstrates in vivo antitumor activity